Assay Details
Target Gene Details
Entrez Gene ID: | 65125 |
Gene Name: | WNK lysine deficient protein kinase 1 |
Gene Aliases: |
HSAN2, HSN2, KDP, PPP1R167, PRKWNK1, PSK, p65 |
Location: |
Chr.12:752552-911452 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2760688 | Chr.12:746699 - 767080 on Build GRCh38 | Loss |
![]() |
esv3548603 | Chr.12:756648 - 766334 on Build GRCh38 | Deletion |
![]() |
nsv556887 | Chr.12:523074 - 758338 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() |
nsv826175 | Chr.12:757211 - 765961 on Build GRCh38 | Loss |
![]() |
nsv8888 | Chr.12:757189 - 765967 on Build GRCh38 | Gain |
![]() |
nsv1119925 | Chr.12:739873 - 764441 on Build GRCh38 | Deletion |
![]() |
nsv1159950 | Chr.12:754908 - 760032 on Build GRCh38 | Duplication |
![]() |
dgv2251n54 | Chr.12:553282 - 1101718 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
dgv431e212 | Chr.12:756526 - 766139 on Build GRCh38 | Loss |
![]() |
dgv2253n54 | Chr.12:741122 - 859323 on Build GRCh38 | Loss |
![]() |
nsv556899 | Chr.12:741447 - 813242 on Build GRCh38 | Loss |
![]() |
dgv1335n100 | Chr.12:739562 - 764851 on Build GRCh38 | Loss |
![]() |
nsv819875 | Chr.12:757124 - 765317 on Build GRCh38 | Gain |
![]() |
esv2759875 | Chr.12:225844 - 881378 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv556893 | Chr.12:628064 - 873155 on Build GRCh38 | Gain |
![]() ![]() ![]() |
nsv518946 | Chr.12:745165 - 852170 on Build GRCh38 | Loss |
![]() |
dgv2256n54 | Chr.12:756804 - 769498 on Build GRCh38 | Loss |
![]() |
nsv1038532 | Chr.12:754667 - 767068 on Build GRCh38 | Gain |
![]() |
nsv556901 | Chr.12:753823 - 760447 on Build GRCh38 | Gain |
![]() |
dgv27e180 | Chr.12:757004 - 766773 on Build GRCh38 | Deletion |
![]() |
esv2870457 | Chr.12:756834 - 766834 on Build GRCh38 | Deletion |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Additional Information:
For this assay, SNP(s) [rs139907131] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)