Assay Details
Target Gene Details
Entrez Gene ID: | 2735 |
Gene Name: | GLI family zinc finger 1 |
Gene Aliases: |
GLI |
Location: |
Chr.12:57460135-57472268 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GLI1 | NM_001160045.1 | NP_001153517.1 | ||
NM_001167609.1 | NP_001161081.1 | |||
NM_005269.2 | NP_005260.1 | |||
XM_011538189.2 | XP_011536491.1 | |||
XM_011538190.1 | XP_011536492.1 | |||
AB239135.1 | ||||
AB239136.1 | ||||
AB239328.1 | ||||
AK297899.1 | ||||
X07384.1 | CAA30297.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv952190 | Chr.12:57459418 - 57466117 on Build GRCh38 | Deletion | GLI1 |
nsv428283 | Chr.12:57435950 - 57608085 on Build GRCh38 | Gain | ARHGAP9 INHBE DCTN2 DDIT3 MIR616 GLI1 MIR6758 INHBC DTX3 PIP4K2C KIF5A MBD6 MARS |
nsv469411 | Chr.12:57450266 - 57527405 on Build GRCh38 | Loss | ARHGAP9 INHBE DDIT3 MIR616 GLI1 MIR6758 MBD6 INHBC MARS |
nsv482992 | Chr.12:56206217 - 57706217 on Build GRCh38 | Gain+Loss | PAN2 LRP1-AS LOC101927583 NDUFA4L2 MYO1A SPRYD4 STAT2 ARHGEF25 STAT6 MIR1228 COQ10A LRP1 NEMP1 OS9 ARHGAP9 RBMS2 DDIT3 MIR616 PRIM1 GLI1 SLC39A5 MIR6758 NACA STAC3 SNORD59A PIP4K2C ZBTB39 NAB2 KIF5A SHMT2 MARS IL23A INHBE DCTN2 SNORD59B NABP2 SDR9C7 PTGES3 INHBC DTX3 GPR182 TIMELESS TAC3 R3HDM2 SNORA105C CNPY2 RDH16 HBCBP GLS2 APOF ATP5B CS BAZ2A SLC26A10 NXPH4 RNF41 MIP MBD6 B4GALNT1 HSD17B6 ANKRD52 |
nsv8974 | Chr.12:57458996 - 57472098 on Build GRCh38 | Gain | GLI1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |