Assay Details
Target Gene Details
Entrez Gene ID: | 100507424 |
Gene Name: | uncharacterized LOC100507424 |
Gene Aliases: |
- |
Location: |
Chr.12:2836816-2859795 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC100507424 | NR_038933.1 | |||
BU947711.1 | ||||
BU947982.1 |
Target Gene Details
Entrez Gene ID: | 101929469 |
Gene Name: | uncharacterized protein ENSP00000372125 |
Gene Aliases: |
- |
Location: |
Chr.12:2849221-2857215 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC101929469 | XM_005253817.3 | XP_005253874.2 | ||
XM_011521043.2 | XP_011519345.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3628321 | Chr.12:2796341 - 2902786 on Build GRCh38 | Gain | ITFG2 LOC101929469 FOXM1 TULP3 NRIP2 RHNO1 LOC100507424 FKBP4 |
dgv19n68 | Chr.12:2726070 - 2935438 on Build GRCh38 | Loss | ITFG2 LOC283440 LOC101929469 FOXM1 TULP3 NRIP2 RHNO1 LOC100507424 FKBP4 |
nsv832311 | Chr.12:2810344 - 2996539 on Build GRCh38 | Loss | ITFG2 TEAD4 LOC101929469 FOXM1 TULP3 NRIP2 RHNO1 LOC100507424 |