Assay Details
Target Gene Details
Entrez Gene ID: | 65125 |
Gene Name: | WNK lysine deficient protein kinase 1 |
Gene Aliases: |
HSAN2, HSN2, KDP, PPP1R167, PRKWNK1, PSK, p65 |
Location: |
Chr.12:752552-911452 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv2251n54 | Chr.12:553282 - 1101718 on Build GRCh38 | Gain | ERC1 NINJ2 LOC100049716 B4GALNT3 WNK1 RAD52 LOC105369595 |
dgv2253n54 | Chr.12:741122 - 859323 on Build GRCh38 | Loss | WNK1 |
nsv556899 | Chr.12:741447 - 813242 on Build GRCh38 | Loss | WNK1 |
esv2760258 | Chr.12:759943 - 776028 on Build GRCh38 | Loss | WNK1 |
esv2759875 | Chr.12:225844 - 881378 on Build GRCh38 | Gain+Loss | NINJ2 LOC100049716 B4GALNT3 WNK1 LOC102723544 KDM5A LOC105369595 CCDC77 SLC6A13 |
nsv556893 | Chr.12:628064 - 873155 on Build GRCh38 | Gain | NINJ2 LOC100049716 WNK1 |
nsv518946 | Chr.12:745165 - 852170 on Build GRCh38 | Loss | WNK1 |
nsv437133 | Chr.12:756804 - 779154 on Build GRCh38 | Loss | WNK1 |
nsv1159952 | Chr.12:764778 - 774012 on Build GRCh38 | Duplication | WNK1 |
nsv952782 | Chr.12:767335 - 814534 on Build GRCh38 | Duplication | WNK1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |