Assay Details
Target Gene Details
Entrez Gene ID: | 2735 |
Gene Name: | GLI family zinc finger 1 |
Gene Aliases: |
GLI |
Location: |
Chr.12:57460135-57472268 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GLI1 | NM_001160045.1 | NP_001153517.1 | ||
NM_001167609.1 | NP_001161081.1 | |||
NM_005269.2 | NP_005260.1 | |||
XM_011538189.2 | XP_011536491.1 | |||
XM_011538190.1 | XP_011536492.1 | |||
AB239135.1 | ||||
AB239136.1 | ||||
AB239328.1 | ||||
AK297899.1 | ||||
X07384.1 | CAA30297.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv952190 | Chr.12:57459418 - 57466117 on Build GRCh38 | Deletion | GLI1 |
nsv428283 | Chr.12:57435950 - 57608085 on Build GRCh38 | Gain | INHBE MIR616 DTX3 MARS DCTN2 MBD6 GLI1 KIF5A INHBC PIP4K2C MIR6758 DDIT3 ARHGAP9 |
nsv469411 | Chr.12:57450266 - 57527405 on Build GRCh38 | Loss | INHBE INHBC MIR616 MARS MIR6758 DDIT3 ARHGAP9 MBD6 GLI1 |
nsv482992 | Chr.12:56206217 - 57706217 on Build GRCh38 | Gain+Loss | MIR616 NABP2 IL23A OS9 ARHGEF25 SLC26A10 TAC3 MBD6 R3HDM2 LOC101927583 APOF NEMP1 ARHGAP9 STAT2 ANKRD52 ZBTB39 INHBE PAN2 SDR9C7 MARS DCTN2 GLI1 HSD17B6 STAT6 SHMT2 INHBC SPRYD4 LRP1 DDIT3 HBCBP BAZ2A SNORD59B COQ10A PRIM1 NAB2 DTX3 MYO1A PTGES3 RBMS2 MIP PIP4K2C STAC3 SNORD59A MIR6758 RDH16 MIR1228 GLS2 CNPY2 NXPH4 CS NDUFA4L2 GPR182 RNF41 B4GALNT1 ATP5B KIF5A NACA LRP1-AS SNORA105C SLC39A5 TIMELESS |
nsv8974 | Chr.12:57458996 - 57472098 on Build GRCh38 | Gain | GLI1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |