Assay Details
Target Gene Details
Entrez Gene ID: | 121256 |
Gene Name: | transmembrane protein 132D |
Gene Aliases: |
MOLT, PPP1R153 |
Location: |
Chr.12:129071726-129903667 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TMEM132D | NM_133448.2 | NP_597705.2 | ||
AB061814.1 | BAC97794.1 | |||
AB075824.1 | BAB85530.1 | |||
BC114626.1 | ||||
BC114936.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv930 | Chr.12:129065704 - 129096684 on Build GRCh38 | Deletion | TMEM132D |
esv3631156 | Chr.12:128909343 - 129187331 on Build GRCh38 | Gain | LOC283352 TMEM132D GLT1D1 |
nsv1040831 | Chr.12:128749443 - 129516029 on Build GRCh38 | Gain | LOC283352 TMEM132D LOC101927735 SLC15A4 LOC100128276 LOC101929984 GLT1D1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |