Assay Details
Target Gene Details
Entrez Gene ID: | 144347 |
Gene Name: | family with sequence similarity 101 member A |
Gene Aliases: |
CFM2 |
Location: |
Chr.12:124289164-124316024 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FAM101A | NM_181709.4 | NP_859060.3 | ||
AK074545.1 | BAC11049.1 | |||
AK126577.1 | BAC86601.1 |
Target Gene Details
Entrez Gene ID: | 105370045 |
Gene Name: | filamin-interacting protein FAM101A |
Gene Aliases: |
- |
Location: |
Chr.12:124295091-124301978 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC105370045 | XM_011539088.1 | XP_011537390.1 |
Target Gene Details
Entrez Gene ID: | 100533183 |
Gene Name: | filamin-interacting protein FAM101A |
Gene Aliases: |
- |
Location: |
Chr.12:123973215-124316024 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ZNF664-FAM101A | NM_001204299.1 | NP_001191228.1 | ||
DA169732.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv818930 | Chr.12:124256444 - 124391587 on Build GRCh38 | Gain | NCOR2 FAM101A LOC105370045 MIR6880 ZNF664-FAM101A |
esv3631012 | Chr.12:124290695 - 124349764 on Build GRCh38 | Gain | NCOR2 FAM101A LOC105370045 MIR6880 ZNF664-FAM101A |
esv3631010 | Chr.12:124288701 - 124749461 on Build GRCh38 | Gain | NCOR2 FAM101A LOC105370045 MIR6880 ZNF664-FAM101A |
nsv952500 | Chr.12:124296455 - 124567454 on Build GRCh38 | Deletion | NCOR2 FAM101A LOC105370045 MIR6880 ZNF664-FAM101A |
esv3631011 | Chr.12:124289768 - 124548418 on Build GRCh38 | Gain | NCOR2 FAM101A LOC105370045 MIR6880 ZNF664-FAM101A |
nsv560539 | Chr.12:124250181 - 124314841 on Build GRCh38 | Loss | FAM101A LOC105370045 ZNF664-FAM101A |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |