Assay Details
Target Gene Details
Entrez Gene ID: | 65125 |
Gene Name: | WNK lysine deficient protein kinase 1 |
Gene Aliases: |
HSAN2, HSN2, KDP, PPP1R167, PRKWNK1, PSK, p65 |
Location: |
Chr.12:752552-911452 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv2251n54 | Chr.12:553282 - 1101718 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
dgv2253n54 | Chr.12:741122 - 859323 on Build GRCh38 | Loss |
![]() |
nsv556899 | Chr.12:741447 - 813242 on Build GRCh38 | Loss |
![]() |
esv2760258 | Chr.12:759943 - 776028 on Build GRCh38 | Loss |
![]() |
esv2759875 | Chr.12:225844 - 881378 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv556893 | Chr.12:628064 - 873155 on Build GRCh38 | Gain |
![]() ![]() ![]() |
nsv518946 | Chr.12:745165 - 852170 on Build GRCh38 | Loss |
![]() |
nsv437133 | Chr.12:756804 - 779154 on Build GRCh38 | Loss |
![]() |
nsv952782 | Chr.12:767335 - 814534 on Build GRCh38 | Duplication |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)