Assay Details
Target Gene Details
Entrez Gene ID: | 57701 |
Gene Name: | NCK associated protein 5 like |
Gene Aliases: |
Cep169, FP1193, KIAA1602 |
Location: |
Chr.12:49791139-49828750 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NCKAP5L | NM_001037806.3 | NP_001032895.2 | ||
XM_005269050.3 | XP_005269107.1 | |||
XM_006719525.2 | XP_006719588.1 | |||
XM_011538610.2 | XP_011536912.1 | |||
XM_011538611.2 | XP_011536913.1 | |||
CN359941.1 | ||||
CN359946.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |