Assay Details
Target Gene Details
Entrez Gene ID: | 6433 |
Gene Name: | splicing factor SWAP homolog |
Gene Aliases: |
SFRS8, SWAP |
Location: |
Chr.12:131711087-131799738 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SFSWAP | NM_001261411.1 | NP_001248340.1 | ||
NM_004592.3 | NP_004583.2 | |||
XM_011538653.1 | XP_011536955.1 | |||
XM_011538654.2 | XP_011536956.1 | |||
XM_011538655.2 | XP_011536957.1 | |||
XM_017019798.1 | XP_016875287.1 | |||
XM_017019799.1 | XP_016875288.1 | |||
AB208921.1 | BAD92158.1 | |||
BC008707.2 | AAH08707.1 | |||
BC015953.2 | AAH15953.1 | |||
BC024041.1 | AAH24041.1 | |||
BC136678.1 | ||||
BC144364.1 | ||||
U08377.1 | AAA19604.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv34554 | Chr.12:131284620 - 131807802 on Build GRCh38 | Gain | LOC100128002 LOC107161159 LOC100996701 SFSWAP LOC338797 |
esv2761778 | Chr.12:131668187 - 131735245 on Build GRCh38 | Gain | SFSWAP |
nsv818946 | Chr.12:131631122 - 131760256 on Build GRCh38 | Gain | LOC100996701 SFSWAP |
nsv1047271 | Chr.12:131597710 - 131742961 on Build GRCh38 | Gain | LOC100996701 SFSWAP |
esv2759922 | Chr.12:131238746 - 131784349 on Build GRCh38 | Gain+Loss | LOC100128002 LOC107161159 LOC100996701 SFSWAP LOC338797 |
nsv826560 | Chr.12:131695304 - 131761272 on Build GRCh38 | Gain | SFSWAP |
dgv1588n100 | Chr.12:131696755 - 131753633 on Build GRCh38 | Gain | SFSWAP |
nsv9044 | Chr.12:131683504 - 131763120 on Build GRCh38 | Gain | SFSWAP |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |