Assay Details
Target Gene Details
Entrez Gene ID: | 283232 |
Gene Name: | transmembrane protein 80 |
Gene Aliases: |
- |
Location: |
Chr.11:695386-705028 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TMEM80 | NM_001042463.2 | NP_001035928.2 | ||
NM_001276253.1 | NP_001263182.1 | |||
NM_001276274.1 | NP_001263203.1 | |||
NM_174940.3 | NP_777600.3 | |||
XM_006718206.3 | XP_006718269.3 | |||
XM_017017600.1 | XP_016873089.1 | |||
AK124309.1 | ||||
AK302585.1 | ||||
BC008671.1 | ||||
BI916171.1 | ||||
CK819789.1 | ||||
DR006597.1 | ||||
HY080028.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3625074 | Chr.11:669311 - 702135 on Build GRCh38 | Loss | TMEM80 DEAF1 |
nsv469923 | Chr.11:378188 - 1302334 on Build GRCh38 | Loss | LOC101927503 PTDSS2 MUC5AC CHID1 MUC5B POLR2L TMEM80 SCT MIR6744 RPLP2 B4GALNT4 LRRC56 PHRF1 ANO9 PDDC1 MUC2 CRACR2B RASSF7 LOC143666 PKP3 DRD4 CEND1 MIR210HG PNPLA2 TALDO1 IRF7 PANO1 MUC6 TSPAN4 SNORA52 HRAS CDHR5 SLC25A22 LOC171391 DEAF1 PIDD1 CD151 LMNTD2 RNH1 AP2A2 SIGIRR TOLLIP LOC101059906 MIR210 EPS8L2 |
nsv832043 | Chr.11:631027 - 758602 on Build GRCh38 | Loss | TALDO1 TMEM80 EPS8L2 DRD4 DEAF1 |
esv2759794 | Chr.11:409846 - 1336162 on Build GRCh38 | Loss | LOC101927503 PTDSS2 MUC5AC CHID1 MUC5B POLR2L TMEM80 SCT MIR6744 RPLP2 LRRC56 PHRF1 ANO9 PDDC1 MUC2 CRACR2B RASSF7 LOC143666 DRD4 CEND1 MIR210HG PNPLA2 TALDO1 IRF7 PANO1 MUC6 TSPAN4 SNORA52 HRAS CDHR5 SLC25A22 LOC171391 DEAF1 PIDD1 CD151 LMNTD2 RNH1 AP2A2 SIGIRR TOLLIP LOC101059906 MIR210 EPS8L2 TOLLIP-AS1 |
dgv182e199 | Chr.11:349830 - 943372 on Build GRCh38 | Deletion | PTDSS2 CHID1 POLR2L TMEM80 SCT RPLP2 B4GALNT4 LRRC56 PHRF1 ANO9 PDDC1 CRACR2B RASSF7 LOC143666 PKP3 DRD4 CEND1 MIR210HG PNPLA2 TALDO1 IRF7 PANO1 TSPAN4 SNORA52 HRAS CDHR5 SLC25A22 LOC171391 DEAF1 PIDD1 CD151 LMNTD2 RNH1 AP2A2 SIGIRR LOC101059906 MIR210 EPS8L2 |
nsv1159791 | Chr.11:461793 - 909492 on Build GRCh38 | Deletion | PTDSS2 CHID1 POLR2L TMEM80 SCT RPLP2 LRRC56 PHRF1 PDDC1 CRACR2B RASSF7 LOC143666 DRD4 CEND1 MIR210HG PNPLA2 TALDO1 IRF7 PANO1 TSPAN4 SNORA52 HRAS CDHR5 SLC25A22 LOC171391 DEAF1 PIDD1 CD151 LMNTD2 RNH1 LOC101059906 MIR210 EPS8L2 |