Assay Details
Target Gene Details
Entrez Gene ID: | 283219 |
Gene Name: | potassium channel tetramerization domain containing 21 |
Gene Aliases: |
KCASH2 |
Location: |
Chr.11:78171243-78188854 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KCTD21 | NM_001029859.2 | NP_001025030.1 | ||
XM_005273925.3 | XP_005273982.1 | |||
XM_005273928.1 | XP_005273985.1 | |||
XM_006718517.2 | XP_006718580.1 | |||
XM_006718518.3 | XP_006718581.1 | |||
XM_011544955.2 | XP_011543257.1 | |||
AK095233.1 | ||||
AK300320.1 | ||||
BC036058.1 | AAH36058.1 | |||
DC342183.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv520740 | Chr.11:77842592 - 78286890 on Build GRCh38 | Gain | NDUFC2-KCTD14 KCTD21-AS1 INTS4 ALG8 GAB2 AAMDC THRSP RSF1 USP35 NDUFC2 KCTD14 KCTD21 |
nsv516503 | Chr.11:78145131 - 78209884 on Build GRCh38 | Loss | KCTD21-AS1 USP35 KCTD21 |
nsv527066 | Chr.11:78172809 - 78252087 on Build GRCh38 | Gain | KCTD21-AS1 GAB2 USP35 KCTD21 |
esv33345 | Chr.11:78177662 - 78190142 on Build GRCh38 | Gain | USP35 KCTD21 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |