Assay Details
Target Gene Details
Entrez Gene ID: | 710 |
Gene Name: | serpin family G member 1 |
Gene Aliases: |
C1IN, C1INH, C1NH, HAE1, HAE2 |
Location: |
Chr.11:57597554-57614853 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SERPING1 | NM_000062.2 | NP_000053.2 | ||
NM_001032295.1 | NP_001027466.1 | |||
AB209826.1 | BAD93063.1 | |||
AK293054.1 | ||||
AK303809.1 | ||||
AK303840.1 | ||||
AK312626.1 | ||||
AY732485.1 | AAV33124.1 | |||
AY732486.1 | AAV33125.1 | |||
BC011171.2 | AAH11171.1 | |||
BT006966.1 | AAP35612.1 | |||
GU727623.1 | ||||
KU177948.1 | ||||
KU177949.1 | ||||
KU177950.1 | ||||
KU177951.1 | ||||
M13656.1 | AAB59387.1 | |||
M13690.1 | AAA35613.1 | |||
M14036.1 | AAA51848.1 | |||
X07577.1 | CAA30469.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv832163 | Chr.11:57531881 - 57713578 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)