Assay Details
Target Gene Details
Entrez Gene ID: | 9866 |
Gene Name: | tripartite motif containing 66 |
Gene Aliases: |
C11orf29, TIF1D, TIF1DELTA |
Location: |
Chr.11:8612037-8683217 on Build GRCh38 |
Assay Gene Location: | Within Exon 33 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2751035 | Chr.11:8410107 - 8663904 on Build GRCh38 | Loss | STK33 TRIM66 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |