Assay Details
Target Gene Details
Entrez Gene ID: | 10077 |
Gene Name: | tetraspanin 32 |
Gene Aliases: |
ART1, PHEMX, PHMX, TSSC6 |
Location: |
Chr.11:2299480-2318201 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TSPAN32 | NM_139022.2 | NP_620591.3 | ||
XM_005252719.4 | XP_005252776.1 | |||
XM_005252720.4 | XP_005252777.1 | |||
XM_011519815.2 | XP_011518117.2 | |||
XM_011519818.2 | XP_011518120.2 | |||
XM_011519819.1 | XP_011518121.1 | |||
XM_011519822.1 | XP_011518124.1 | |||
XM_011519828.2 | XP_011518130.1 | |||
XM_011519829.2 | XP_011518131.2 | |||
XM_017017067.1 | XP_016872556.1 | |||
AF125569.1 | AAD23580.1 | |||
AF176070.1 | AAG15840.1 | |||
AF176071.1 | AAG15841.1 | |||
AK128812.1 | ||||
AK298692.1 | ||||
AK307638.1 | ||||
AK310116.1 | ||||
AY039001.1 | AAK84431.1 | |||
AY303780.1 | AAP76185.1 | |||
BC016693.1 | AAH16693.1 | |||
BG506933.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3625125 | Chr.11:2303216 - 2328448 on Build GRCh38 | Gain | TSPAN32 |
nsv469928 | Chr.11:2294174 - 2473462 on Build GRCh38 | Loss | CD81-AS1 CD81 KCNQ1 C11orf21 TSPAN32 TRPM5 TSSC4 |
nsv553081 | Chr.11:2291007 - 2312445 on Build GRCh38 | Loss | C11orf21 TSPAN32 |
nsv951285 | Chr.11:2256071 - 2310670 on Build GRCh38 | Deletion | C11orf21 TSPAN32 ASCL2 |
nsv553047 | Chr.11:1733170 - 2418537 on Build GRCh38 | Gain | CD81 SYT8 C11orf21 IGF2 HOTS MIR4686 LINC01150 TH TNNT3 INS-IGF2 IGF2-AS TSPAN32 MIR4298 H19 TRPM5 TSSC4 MRPL23 MIR675 CD81-AS1 MIR483 LSP1 CTSD INS MRPL23-AS1 TNNI2 IFITM10 SNORD131 LINC01219 MIR7847 ASCL2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |