Assay Details
Target Gene Details
Entrez Gene ID: | 26610 |
Gene Name: | elongator acetyltransferase complex subunit 4 |
Gene Aliases: |
AN, C11orf19, PAX6NEB, PAXNEB, dJ68P15A.1, hELP4 |
Location: |
Chr.11:31509729-31784525 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ELP4 | NM_001288725.1 | NP_001275654.1 | ||
NM_001288726.1 | NP_001275655.1 | |||
NM_019040.4 | NP_061913.3 | |||
AK000505.1 | BAA91212.1 | |||
AK304885.1 | ||||
AL136677.1 | CAB66612.1 | |||
BC012514.1 | AAH12514.1 | |||
HY127287.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv467785 | Chr.11:31467343 - 31674788 on Build GRCh38 | Loss |
![]() ![]() |
dgv1740n54 | Chr.11:31433428 - 31674788 on Build GRCh38 | Loss |
![]() ![]() |
esv2751022 | Chr.11:31041408 - 31526385 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() |
esv3625830 | Chr.11:31513026 - 31809558 on Build GRCh38 | Gain |
![]() ![]() |
nsv529004 | Chr.11:30992197 - 31700936 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() |
nsv1052451 | Chr.11:31495114 - 31533123 on Build GRCh38 | Loss |
![]() ![]() |
nsv553988 | Chr.11:31513808 - 31629642 on Build GRCh38 | Loss |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)