Assay Details
Target Gene Details
Entrez Gene ID: | 25906 |
Gene Name: | anaphase promoting complex subunit 15 |
Gene Aliases: |
APC15, C11orf51, HSPC020 |
Location: |
Chr.11:72106254-72112793 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ANAPC15 | XM_017017488.1 | XP_016872977.1 | ||
XM_017017489.1 | XP_016872978.1 | |||
XM_017017490.1 | XP_016872979.1 | |||
XM_017017491.1 | XP_016872980.1 | |||
XM_017017492.1 | XP_016872981.1 | |||
XM_017017493.1 | XP_016872982.1 | |||
XM_017017494.1 | XP_016872983.1 | |||
XM_017017495.1 | XP_016872984.1 | |||
AK311146.1 |
Target Gene Details
Entrez Gene ID: | 220074 |
Gene Name: | leucine rich transmembrane and O-methyltransferase domain containing |
Gene Aliases: |
CFAP111, DFNB63, LRRC51 |
Location: |
Chr.11:72079982-72110782 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LRTOMT | NM_001145308.4 | 7 | 1672 | NP_001138780.1 |
NM_001145309.3 | 9 | 1893 | NP_001138781.1 | |
NM_001145310.3 | 9 | 1773 | NP_001138782.1 | |
XM_017017356.1 | 4 | 2287 | XP_016872845.1 | |
XM_017017357.1 | 3 | 3210 | XP_016872846.1 | |
XM_017017358.1 | 3 | 3210 | XP_016872847.1 | |
AF289588.1 | 5 | 754 | AAL55772.1 | |
AK302772.1 | 7 | 1166 | ||
BX115135.1 | 1 | 181 | ||
EU627069.1 | 9 | 1365 | ||
EU627070.1 | 9 | 1245 |
More Information
Set Membership: |
Intragenic Exonic Intronic Non-exonic non-DGV Variation |