Assay Details
Target Gene Details
Entrez Gene ID: | 161 |
Gene Name: | adaptor related protein complex 2 alpha 2 subunit |
Gene Aliases: |
ADTAB, CLAPA2, HIP-9, HIP9, HYPJ |
Location: |
Chr.11:925809-1012245 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AP2A2 | NM_001242837.1 | NP_001229766.1 | ||
NM_012305.3 | NP_036437.1 | |||
XM_011519929.1 | XP_011518231.1 | |||
XM_011519930.2 | XP_011518232.1 | |||
AB020706.2 | BAA74922.2 | |||
AK001986.1 | ||||
AK027891.1 | BAB55435.1 | |||
AK223558.1 | BAD97278.1 | |||
AK293763.1 | ||||
AK299364.1 | ||||
BC006155.1 | AAH06155.1 | |||
BG705558.1 | ||||
BI462785.1 | ||||
DB461570.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3625088 | Chr.11:876861 - 979340 on Build GRCh38 | Gain | CHID1 AP2A2 |
nsv469923 | Chr.11:378188 - 1302334 on Build GRCh38 | Loss | MIR6744 SLC25A22 TMEM80 PKP3 B4GALNT4 IRF7 MUC5AC MIR210 CD151 LOC101927503 TOLLIP CHID1 MUC6 LOC143666 PHRF1 SNORA52 TALDO1 EPS8L2 SIGIRR DRD4 LOC171391 RASSF7 PANO1 MUC5B CEND1 POLR2L AP2A2 PIDD1 TSPAN4 LMNTD2 PTDSS2 SCT HRAS CRACR2B DEAF1 LOC101059906 CDHR5 RPLP2 LRRC56 PNPLA2 RNH1 ANO9 PDDC1 MUC2 MIR210HG |
dgv175e214 | Chr.11:851409 - 1043855 on Build GRCh38 | Gain | CHID1 MUC6 AP2A2 TSPAN4 |
nsv428244 | Chr.11:882364 - 1061595 on Build GRCh38 | Loss | CHID1 MUC6 AP2A2 LOC101927503 |
esv2759794 | Chr.11:409846 - 1336162 on Build GRCh38 | Loss | MIR6744 SLC25A22 TMEM80 IRF7 MUC5AC MIR210 CD151 LOC101927503 TOLLIP CHID1 MUC6 LOC143666 PHRF1 SNORA52 TALDO1 EPS8L2 SIGIRR DRD4 LOC171391 RASSF7 PANO1 MUC5B TOLLIP-AS1 CEND1 POLR2L AP2A2 PIDD1 TSPAN4 LMNTD2 PTDSS2 SCT HRAS CRACR2B DEAF1 LOC101059906 CDHR5 RPLP2 LRRC56 PNPLA2 RNH1 ANO9 PDDC1 MUC2 MIR210HG |
esv2669189 | Chr.11:864834 - 1489637 on Build GRCh38 | Deletion | MIR6744 AP2A2 TSPAN4 MUC5AC LOC101927503 MOB2 TOLLIP CHID1 MUC6 BRSK2 MUC2 MUC5B TOLLIP-AS1 |
dgv182e199 | Chr.11:349830 - 943372 on Build GRCh38 | Deletion | SLC25A22 TMEM80 PKP3 B4GALNT4 IRF7 MIR210 CD151 CHID1 LOC143666 PHRF1 SNORA52 TALDO1 EPS8L2 SIGIRR DRD4 LOC171391 RASSF7 PANO1 CEND1 POLR2L AP2A2 PIDD1 TSPAN4 LMNTD2 PTDSS2 SCT HRAS CRACR2B DEAF1 LOC101059906 CDHR5 RPLP2 LRRC56 PNPLA2 RNH1 ANO9 PDDC1 MIR210HG |
esv2743878 | Chr.11:864732 - 1489652 on Build GRCh38 | Deletion | MIR6744 AP2A2 TSPAN4 MUC5AC LOC101927503 MOB2 TOLLIP CHID1 MUC6 BRSK2 MUC2 MUC5B TOLLIP-AS1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |