Assay Details
Target Gene Details
Entrez Gene ID: | 26472 |
Gene Name: | protein phosphatase 1 regulatory inhibitor subunit 14B |
Gene Aliases: |
PHI-1, PLCB3N, PNG, SOM172 |
Location: |
Chr.11:64244479-64246941 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PPP1R14B | NM_138689.2 | NP_619634.1 | ||
AF318364.1 | 1 | 212 | AAL55871.1 | |
BC014522.2 | AAH14522.3 | |||
BC094817.1 | AAH94817.1 | |||
BF155230.1 | ||||
BT009914.1 | AAP88916.1 | |||
X91195.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1159904 | Chr.11:64150129 - 64279169 on Build GRCh38 | Deletion | NUDT22 TRPT1 PLCB3 MACROD1 GPR137 FERMT3 BAD FKBP2 PPP1R14B DNAJC4 VEGFB LOC105369340 STIP1 |
nsv951010 | Chr.11:63942829 - 64352128 on Build GRCh38 | Deletion | NUDT22 MIR7155 PLCB3 MACROD1 GPR137 FERMT3 ESRRA OTUB1 PPP1R14B DNAJC4 LOC105369340 STIP1 COX8A TRPT1 CCDC88B TEX40 NAA40 PRDX5 BAD FLRT1 FKBP2 TRMT112 KCNK4-TEX40 KCNK4 VEGFB |
dgv1963n54 | Chr.11:64174262 - 64329761 on Build GRCh38 | Loss | NUDT22 TRPT1 TEX40 PLCB3 GPR137 FERMT3 PRDX5 BAD FKBP2 ESRRA TRMT112 PPP1R14B KCNK4-TEX40 DNAJC4 KCNK4 VEGFB LOC105369340 STIP1 |
nsv555187 | Chr.11:64185649 - 64260416 on Build GRCh38 | Loss | FERMT3 NUDT22 TRPT1 FKBP2 PPP1R14B DNAJC4 VEGFB LOC105369340 STIP1 PLCB3 |
nsv983032 | Chr.11:64243929 - 64247666 on Build GRCh38 | Duplication | FKBP2 PPP1R14B LOC105369340 |
esv24861 | Chr.11:64233793 - 64250642 on Build GRCh38 | Gain | FKBP2 PPP1R14B DNAJC4 VEGFB LOC105369340 |
nsv832186 | Chr.11:64204788 - 64402587 on Build GRCh38 | Loss | NUDT22 MIR7155 TRPT1 CCDC88B RPS6KA4 TEX40 PLCB3 MIR1237 GPR137 FERMT3 PRDX5 BAD FKBP2 ESRRA TRMT112 PPP1R14B KCNK4-TEX40 DNAJC4 KCNK4 VEGFB LOC105369340 |
nsv555184 | Chr.11:64156974 - 64275525 on Build GRCh38 | Loss | NUDT22 TRPT1 PLCB3 MACROD1 GPR137 FERMT3 BAD FKBP2 PPP1R14B DNAJC4 VEGFB LOC105369340 STIP1 |
nsv469961 | Chr.11:64055468 - 64318495 on Build GRCh38 | Loss | NUDT22 TRPT1 TEX40 PLCB3 MACROD1 GPR137 FERMT3 PRDX5 BAD FLRT1 FKBP2 ESRRA TRMT112 PPP1R14B KCNK4-TEX40 DNAJC4 KCNK4 VEGFB LOC105369340 STIP1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |