Assay Details
Target Gene Details
Entrez Gene ID: | 57558 |
Gene Name: | ubiquitin specific peptidase 35 |
Gene Aliases: |
- |
Location: |
Chr.11:78188812-78233651 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
USP35 | NM_020798.2 | NP_065849.1 | ||
XM_011545187.2 | XP_011543489.1 | |||
XM_011545188.2 | XP_011543490.1 | |||
XM_017018052.1 | XP_016873541.1 | |||
XM_017018053.1 | XP_016873542.1 | |||
AB037793.1 | BAA92610.1 | |||
AJ586137.1 | CAE51937.1 | |||
AK056231.1 | ||||
AK301048.1 | ||||
AK307875.1 | ||||
BC131489.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv520740 | Chr.11:77842592 - 78286890 on Build GRCh38 | Gain | NDUFC2-KCTD14 INTS4 USP35 GAB2 ALG8 RSF1 AAMDC NDUFC2 KCTD21 KCTD21-AS1 KCTD14 THRSP |
nsv516503 | Chr.11:78145131 - 78209884 on Build GRCh38 | Loss | USP35 KCTD21 KCTD21-AS1 |
nsv1036793 | Chr.11:78198402 - 78265821 on Build GRCh38 | Gain | USP35 GAB2 |
nsv527066 | Chr.11:78172809 - 78252087 on Build GRCh38 | Gain | USP35 GAB2 KCTD21 KCTD21-AS1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |