Assay Details
Target Gene Details
Entrez Gene ID: | 22997 |
Gene Name: | immunoglobulin superfamily member 9B |
Gene Aliases: |
- |
Location: |
Chr.11:133908564-133956960 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
IGSF9B | NM_001277285.1 | NP_001264214.1 | ||
XM_011542691.2 | XP_011540993.1 | |||
XM_011542692.2 | XP_011540994.1 | |||
XM_011542694.2 | XP_011540996.1 | |||
XM_011542695.2 | XP_011540997.1 | |||
XM_017017393.1 | XP_016872882.1 | |||
XM_017017394.1 | XP_016872883.1 | |||
XM_017017395.1 | XP_016872884.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv826145 | Chr.11:133933178 - 133964787 on Build GRCh38 | Loss | IGSF9B |
nsv1051677 | Chr.11:133862255 - 133955026 on Build GRCh38 | Gain | MIR4697HG MIR4697 IGSF9B |
dgv1301n100 | Chr.11:133922841 - 133959811 on Build GRCh38 | Gain | IGSF9B |
nsv952765 | Chr.11:133891206 - 133952805 on Build GRCh38 | Deletion | MIR4697HG MIR4697 IGSF9B |
nsv826144 | Chr.11:133920301 - 134020350 on Build GRCh38 | Gain | IGSF9B |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |