Assay Details
Target Gene Details
Entrez Gene ID: | 144132 |
Gene Name: | dynein heavy chain domain 1 |
Gene Aliases: |
C11orf47, CCDC35, DHCD1, DNHD1L |
Location: |
Chr.11:6497296-6572025 on Build GRCh38 |
Assay Gene Location: | Within Intron 22 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DNHD1 | NM_144666.2 | NP_653267.2 | ||
AB290171.1 | ||||
BX647806.1 |
Target Gene Details
Entrez Gene ID: | 23378 |
Gene Name: | ribosomal RNA processing 8, methyltransferase, homolog (yeast) |
Gene Aliases: |
KIAA0409, NML |
Location: |
Chr.11:6514830-6603678 on Build GRCh38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RRP8 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)