Assay Details
Target Gene Details
Entrez Gene ID: | 23769 |
Gene Name: | fibronectin leucine rich transmembrane protein 1 |
Gene Aliases: |
SPG68 |
Location: |
Chr.11:64035970-64119183 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FLRT1 | XM_005273861.1 | XP_005273918.1 |
Target Gene Details
Entrez Gene ID: | 28992 |
Gene Name: | MACRO domain containing 1 |
Gene Aliases: |
LRP16 |
Location: |
Chr.11:63998554-64166652 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MACROD1 | NM_014067.3 | NP_054786.2 | ||
XM_005273939.4 | XP_005273996.1 | |||
XM_011544970.2 | XP_011543272.1 | |||
AF086206.1 | ||||
AF202922.2 | AAF15294.2 | |||
BC000270.1 | AAH00270.2 | |||
BC003188.1 | AAH03188.1 | |||
BC007297.1 | AAH07297.1 | |||
BC008316.1 | AAH08316.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv354 | Chr.11:64044979 - 64079484 on Build GRCh38 | Insertion | FLRT1 MACROD1 |
nsv951010 | Chr.11:63942829 - 64352128 on Build GRCh38 | Deletion | VEGFB TEX40 PRDX5 STIP1 NAA40 LOC105369340 CCDC88B ESRRA NUDT22 FKBP2 KCNK4 FLRT1 MACROD1 FERMT3 KCNK4-TEX40 PLCB3 COX8A GPR137 PPP1R14B OTUB1 MIR7155 DNAJC4 TRPT1 BAD TRMT112 |
esv29993 | Chr.11:63971170 - 64096739 on Build GRCh38 | Loss | FLRT1 COX8A MACROD1 OTUB1 |
nsv555182 | Chr.11:64019917 - 64082340 on Build GRCh38 | Loss | FLRT1 MACROD1 |
dgv15n68 | Chr.11:64020230 - 64196060 on Build GRCh38 | Loss | FLRT1 MACROD1 STIP1 |
nsv555183 | Chr.11:64039521 - 64077211 on Build GRCh38 | Loss | FLRT1 MACROD1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |