Assay Details
Target Gene Details
Entrez Gene ID: | 219844 |
Gene Name: | HYLS1, centriolar and ciliogenesis associated |
Gene Aliases: |
HLS |
Location: |
Chr.11:125883614-125900646 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HYLS1 | NM_145014.2 | NP_659451.1 | ||
XM_011542659.2 | XP_011540961.1 | |||
AK057477.1 | BAB71503.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv248n67 | Chr.11:125843561 - 125919531 on Build GRCh38 | Gain | DDX25 PUS3 HYLS1 LOC105369553 |
nsv556526 | Chr.11:125778321 - 125896149 on Build GRCh38 | Loss | PATE3 PATE2 PATE4 PUS3 HYLS1 LOC105369553 |
nsv556525 | Chr.11:125690285 - 125971104 on Build GRCh38 | Gain | DDX25 PATE3 LOC338667 CDON PATE2 PATE4 PATE1 PUS3 HYLS1 LOC105369553 |
esv32964 | Chr.11:125495069 - 126046387 on Build GRCh38 | Gain+Loss | PATE3 CDON PATE4 PATE1 PUS3 FEZ1 HYLS1 EI24 LOC403312 DDX25 LOC338667 PATE2 ACRV1 CHEK1 STT3A LOC105369553 STT3A-AS1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |