Assay Details
Target Gene Details
Entrez Gene ID: | 9049 |
Gene Name: | aryl hydrocarbon receptor interacting protein |
Gene Aliases: |
ARA9, FKBP16, FKBP37, SMTPHN, XAP-2, XAP2 |
Location: |
Chr.11:67483029-67491108 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AIP | NM_001302959.1 | NP_001289888.1 | ||
NM_001302960.1 | NP_001289889.1 | |||
NM_003977.3 | NP_003968.3 | |||
BC104797.1 | AAI04798.1 | |||
BC104827.1 | AAI04828.1 | |||
BG821308.1 | ||||
BQ054206.1 | ||||
HY065637.1 | ||||
U31913.1 | AAB39923.1 | |||
U78521.1 | AAB59004.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv468604 | Chr.11:67418529 - 67516420 on Build GRCh38 | Loss | CARNS1 RPS6KB2 CDK2AP2 TMEM134 GPR152 PTPRCAP PITPNM1 AIP MIR6752 CORO1B CABP4 |
esv2422195 | Chr.11:67466851 - 67859430 on Build GRCh38 | Duplication | FAM86C2P ACY3 TBX10 CDK2AP2 C11orf72 NUDT8 CABP2 AIP NDUFV1 MIR6752 GSTP1 TMEM134 ALDH3B2 DOC2GP PITPNM1 |
dgv1977n54 | Chr.11:67397024 - 67521123 on Build GRCh38 | Loss | RPS6KB2 CDK2AP2 CABP2 AIP MIR6752 CABP4 CARNS1 TMEM134 RAD9A GPR152 PTPRCAP PITPNM1 TBC1D10C CORO1B PPP1CA |
nsv368 | Chr.11:67479842 - 67509866 on Build GRCh38 | Insertion | CDK2AP2 PITPNM1 AIP MIR6752 |
dgv52n111 | Chr.11:67362651 - 67521123 on Build GRCh38 | Deletion | RPS6KB2 CDK2AP2 CABP2 AIP MIR6752 CABP4 LOC100130987 CARNS1 TMEM134 CLCF1 RAD9A GPR152 PTPRCAP PITPNM1 TBC1D10C CORO1B PPP1CA |
nsv521723 | Chr.11:67408221 - 67543787 on Build GRCh38 | Loss | RPS6KB2 CDK2AP2 CABP2 AIP MIR6752 CABP4 CARNS1 TMEM134 GPR152 PTPRCAP PITPNM1 TBC1D10C CORO1B |
nsv527056 | Chr.11:67439955 - 67526837 on Build GRCh38 | Loss | CDK2AP2 TMEM134 GPR152 PITPNM1 CABP2 AIP MIR6752 CORO1B CABP4 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |