Assay Details
Target Gene Details
Entrez Gene ID: | 64787 |
Gene Name: | EPS8 like 2 |
Gene Aliases: |
EPS8R2 |
Location: |
Chr.11:705217-727727 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EPS8L2 | XM_017018130.1 | 1 | 141 | XP_016873619.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv522733 | Chr.11:702250 - 752059 on Build GRCh38 | Loss | TALDO1 EPS8L2 TMEM80 |
nsv469923 | Chr.11:378188 - 1302334 on Build GRCh38 | Loss | MIR6744 MIR210 TOLLIP MUC5AC LOC101927503 CRACR2B PANO1 PTDSS2 PDDC1 LOC171391 SNORA52 TSPAN4 DRD4 MIR210HG PNPLA2 RPLP2 LMNTD2 EPS8L2 CEND1 CDHR5 RNH1 IRF7 DEAF1 MUC5B LRRC56 AP2A2 SCT MUC6 TMEM80 LOC143666 SLC25A22 RASSF7 B4GALNT4 CD151 SIGIRR ANO9 PIDD1 PKP3 CHID1 TALDO1 POLR2L LOC101059906 PHRF1 MUC2 HRAS |
nsv832043 | Chr.11:631027 - 758602 on Build GRCh38 | Loss | DEAF1 TALDO1 EPS8L2 TMEM80 DRD4 |
esv2759794 | Chr.11:409846 - 1336162 on Build GRCh38 | Loss | MIR6744 MIR210 TOLLIP MUC5AC LOC101927503 CRACR2B PANO1 PTDSS2 TOLLIP-AS1 PDDC1 LOC171391 SNORA52 TSPAN4 DRD4 MIR210HG PNPLA2 RPLP2 LMNTD2 EPS8L2 CEND1 CDHR5 RNH1 IRF7 DEAF1 MUC5B LRRC56 AP2A2 SCT MUC6 TMEM80 LOC143666 SLC25A22 RASSF7 CD151 SIGIRR ANO9 PIDD1 CHID1 TALDO1 POLR2L LOC101059906 PHRF1 MUC2 HRAS |
nsv552869 | Chr.11:702097 - 727446 on Build GRCh38 | Loss | EPS8L2 TMEM80 |
dgv182e199 | Chr.11:349830 - 943372 on Build GRCh38 | Deletion | MIR210 CRACR2B PANO1 PTDSS2 PDDC1 LOC171391 SNORA52 TSPAN4 DRD4 MIR210HG PNPLA2 RPLP2 LMNTD2 EPS8L2 CEND1 CDHR5 RNH1 IRF7 DEAF1 LRRC56 AP2A2 SCT TMEM80 LOC143666 SLC25A22 RASSF7 B4GALNT4 CD151 SIGIRR ANO9 PIDD1 PKP3 CHID1 TALDO1 POLR2L LOC101059906 PHRF1 HRAS |
nsv1159791 | Chr.11:461793 - 909492 on Build GRCh38 | Deletion | MIR210 CRACR2B PANO1 PTDSS2 PDDC1 LOC171391 SNORA52 TSPAN4 DRD4 MIR210HG PNPLA2 RPLP2 LMNTD2 EPS8L2 CEND1 CDHR5 RNH1 IRF7 DEAF1 LRRC56 SCT TMEM80 LOC143666 SLC25A22 RASSF7 CD151 PIDD1 CHID1 TALDO1 POLR2L LOC101059906 PHRF1 HRAS |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |