Assay Details
Target Gene Details
Entrez Gene ID: | 29118 |
Gene Name: | DEAD-box helicase 25 |
Gene Aliases: |
GRTH |
Location: |
Chr.11:125903275-125923110 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DDX25 | NM_013264.4 | NP_037396.3 | ||
XM_011542790.2 | XP_011541092.1 | |||
XM_011542791.2 | XP_011541093.1 | |||
XM_017017626.1 | XP_016873115.1 | |||
AF155140.2 | AAF21371.2 | |||
AK295121.1 | ||||
AK312772.1 | ||||
BC035388.1 | ||||
BC050360.1 | ||||
CD657404.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv535 | Chr.11:125918491 - 125963371 on Build GRCh38 | Deletion |
![]() ![]() ![]() |
nsv556525 | Chr.11:125690285 - 125971104 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv32964 | Chr.11:125495069 - 126046387 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)