Assay Details
Target Gene Details
Entrez Gene ID: | 1662 |
Gene Name: | DEAD-box helicase 10 |
Gene Aliases: |
HRH-J8 |
Location: |
Chr.11:108665025-108940930 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DDX10 | NM_004398.3 | NP_004389.2 | ||
AB040537.1 | BAB18536.1 | |||
AB209884.1 | BAD93121.1 | |||
AK296752.1 | ||||
AK315216.1 | ||||
BC049217.1 | AAH49217.1 | |||
BC091521.1 | AAH91521.1 | |||
BC093654.1 | AAH93654.1 | |||
BC093656.1 | AAH93656.1 | |||
U28042.1 | AAC50823.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3627681 | Chr.11:108607953 - 108849278 on Build GRCh38 | Gain |
![]() |
esv3627682 | Chr.11:108681110 - 108760073 on Build GRCh38 | Gain |
![]() |
nsv832264 | Chr.11:108578886 - 108752993 on Build GRCh38 | Gain+Loss |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)