Assay Details
Target Gene Details
Entrez Gene ID: | 387787 |
Gene Name: | lipoyl(octanoyl) transferase 2 (putative) |
Gene Aliases: |
- |
Location: |
Chr.11:74490727-74493972 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LIPT2 | NM_001144869.1 | 2 | 795 | NP_001138341.1 |
XM_011545021.2 | 2 | 1095 | XP_011543323.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv34979 | Chr.11:74167643 - 74822407 on Build GRCh38 | Gain | MIR4696 PGM2L1 LOC101928580 RNF169 POLD3 P4HA3 C2CD3 LOC100287896 LIPT2 KCNE3 PPME1 CHRDL2 MIR548AL |
nsv555444 | Chr.11:74489694 - 74510125 on Build GRCh38 | Loss | LIPT2 LOC100287896 |
esv2759837 | Chr.11:74131564 - 74846552 on Build GRCh38 | Gain | MIR4696 PGM2L1 LOC101928580 RNF169 POLD3 P4HA3 C2CD3 LOC100287896 LIPT2 KCNE3 PPME1 CHRDL2 XRRA1 MIR548AL |
nsv832209 | Chr.11:74444838 - 74618074 on Build GRCh38 | Gain | LIPT2 KCNE3 POLD3 LOC100287896 |
dgv209e214 | Chr.11:74225600 - 74828943 on Build GRCh38 | Gain | MIR4696 PGM2L1 LIPT2 KCNE3 LOC101928580 PPME1 CHRDL2 RNF169 POLD3 P4HA3 MIR548AL LOC100287896 |
nsv825994 | Chr.11:74225622 - 74823410 on Build GRCh38 | Gain | MIR4696 PGM2L1 LIPT2 KCNE3 LOC101928580 PPME1 CHRDL2 RNF169 POLD3 P4HA3 MIR548AL LOC100287896 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |