Assay Details
Target Gene Details
Entrez Gene ID: | 5592 |
Gene Name: | protein kinase, cGMP-dependent, type I |
Gene Aliases: |
AAT8, PKG, PRKG1B, PRKGR1B, cGK, cGK 1, cGK1, cGKI, cGKI-BETA, cGKI-alpha |
Location: |
Chr.10:50991151-52298350 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PRKG1 | NM_001098512.2 | NP_001091982.1 | ||
AK300110.1 | ||||
D45864.1 | BAA08297.1 | |||
DC310312.1 | ||||
DC379897.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831873 | Chr.10:50950850 - 51139780 on Build GRCh38 | Gain | PRKG1 LOC102724719 |
nsv831872 | Chr.10:50903568 - 51058777 on Build GRCh38 | Gain | PRKG1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |