Assay Details
Target Gene Details
Entrez Gene ID: | 80114 |
Gene Name: | BicC family RNA binding protein 1 |
Gene Aliases: |
BICC, CYSRD |
Location: |
Chr.10:58512970-58831437 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
BICC1 | NM_001080512.2 | NP_001073981.1 | ||
XM_011540185.2 | XP_011538487.1 | |||
AY726586.1 | ||||
CF242850.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv508586 | Chr.10:58516186 - 58621289 on Build GRCh38 | Deletion |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)