Assay Details
Target Gene Details
Entrez Gene ID: | 389932 |
Gene Name: | aldo-keto reductase family 1 member C6, pseudogene |
Gene Aliases: |
TAKR |
Location: |
Chr.10:4871667-4916273 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AKR1C6P | NR_026743.1 | |||
AB037903.1 | BAA92888.1 |
Target Gene Details
Entrez Gene ID: | 83592 |
Gene Name: | aldo-keto reductase family 1 member E2 |
Gene Aliases: |
AKR1CL2, AKRDC1, LoopADR, hTSP, htAKR |
Location: |
Chr.10:4824554-4908421 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AKR1E2 | XM_017016744.1 | XP_016872233.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3622212 | Chr.10:4867721 - 4880131 on Build GRCh38 | Loss | AKR1E2 AKR1C6P |
nsv549881 | Chr.10:4859606 - 5277716 on Build GRCh38 | Loss | AKR1C8P AKR1E2 AKR1C3 AKR1C2 AKR1C6P AKR1C4 AKR1C1 |
nsv1045155 | Chr.10:4830047 - 5169355 on Build GRCh38 | Loss | AKR1C8P AKR1E2 AKR1C3 AKR1C2 AKR1C6P AKR1C1 |
esv2762839 | Chr.10:4874422 - 4886698 on Build GRCh38 | Loss | AKR1E2 AKR1C6P |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |