Assay Details
Target Gene Details
Entrez Gene ID: | 101927824 |
Gene Name: | translation initiation factor IF-2-like |
Gene Aliases: |
- |
Location: |
Chr.10:3052510-3069273 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC101927824 | XM_011519779.1 | XP_011518081.1 | ||
XM_011519781.1 | XP_011518083.1 | |||
XM_017017013.1 | XP_016872502.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv549814 | Chr.10:3055020 - 3170233 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() |
dgv932n54 | Chr.10:2382612 - 3253481 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv2730995 | Chr.10:2770220 - 3706473 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv428229 | Chr.10:2618610 - 3156581 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() |
esv2760124 | Chr.10:2641047 - 3094525 on Build GRCh38 | Gain |
![]() ![]() |
nsv517630 | Chr.10:2632486 - 3097509 on Build GRCh38 | Gain+Loss |
![]() ![]() |
nsv470913 | Chr.10:3052993 - 3125271 on Build GRCh38 | Loss |
![]() ![]() |
nsv549810 | Chr.10:3035272 - 3691194 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv34832 | Chr.10:2638872 - 3091456 on Build GRCh38 | Gain |
![]() ![]() |
dgv933n54 | Chr.10:2644284 - 3091456 on Build GRCh38 | Gain |
![]() ![]() |
nsv470912 | Chr.10:2650464 - 3091456 on Build GRCh38 | Gain |
![]() ![]() |
esv26266 | Chr.10:2646665 - 3092022 on Build GRCh38 | Gain+Loss |
![]() ![]() |
dgv660n100 | Chr.10:2641472 - 3092489 on Build GRCh38 | Gain |
![]() ![]() |
dgv935n54 | Chr.10:3044294 - 3125271 on Build GRCh38 | Loss |
![]() ![]() |
esv3622141 | Chr.10:2646620 - 3092104 on Build GRCh38 | Gain |
![]() ![]() |
nsv825215 | Chr.10:3058658 - 3099236 on Build GRCh38 | Gain |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)