Assay Details
Target Gene Details
Entrez Gene ID: | 283050 |
Gene Name: | ZMIZ1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.10:78943326-79067448 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ZMIZ1-AS1 | NR_015429.1 | |||
NR_024429.1 | ||||
NR_024431.2 | ||||
AK097923.1 | ||||
AK098249.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv551561 | Chr.10:78349364 - 79336832 on Build GRCh38 | Gain | ZMIZ1-AS1 ZMIZ1 |
nsv7343 | Chr.10:78955245 - 79019009 on Build GRCh38 | Insertion | ZMIZ1-AS1 |