Assay Details
Target Gene Details
Entrez Gene ID: | 57715 |
Gene Name: | semaphorin 4G |
Gene Aliases: |
- |
Location: |
Chr.10:100969483-100985618 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SEMA4G | NM_001203244.1 | NP_001190173.1 | ||
NM_017893.3 | NP_060363.2 | |||
XM_005270008.2 | XP_005270065.1 | |||
AB046839.1 | ||||
AK297431.1 | ||||
BC051030.1 | AAH51030.1 | |||
BC110382.1 | ||||
BC128579.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831965 | Chr.10:100916724 - 101082796 on Build GRCh38 | Loss | MRPL43 LZTS2 C10orf2 SEMA4G KAZALD1 MIR608 PDZD7 SLF2 SFXN3 |
esv3624366 | Chr.10:100974364 - 100977231 on Build GRCh38 | Loss | SEMA4G MIR608 |
nsv517698 | Chr.10:100965896 - 101017113 on Build GRCh38 | Loss | MRPL43 LZTS2 C10orf2 SEMA4G MIR608 PDZD7 |
nsv467443 | Chr.10:100929460 - 101007360 on Build GRCh38 | Loss | MRPL43 LZTS2 C10orf2 SEMA4G MIR608 SLF2 |
nsv552051 | Chr.10:100904892 - 100984619 on Build GRCh38 | Loss | MRPL43 SEMA4G MIR608 SLF2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |