Assay Details
Target Gene Details
Entrez Gene ID: | 3778 |
Gene Name: | potassium calcium-activated channel subfamily M alpha 1 |
Gene Aliases: |
BKTM, KCa1.1, MaxiK, SAKCA, SLO, SLO-ALPHA, SLO1, bA205K10.1, hSlo, mSLO1 |
Location: |
Chr.10:76869601-77637819 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Target Gene Details
Entrez Gene ID: | 101929286 |
Gene Name: | KCNMA1 antisense RNA 3 |
Gene Aliases: |
- |
Location: |
Chr.10:77350872-77376613 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KCNMA1-AS3 | NR_126365.1 | |||
AA454124.1 | ||||
AW510993.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2759769 | Chr.10:77351633 - 77833713 on Build GRCh38 | Gain+Loss | KCNMA1 DLG5 KCNMA1-AS3 |
nsv551551 | Chr.10:77261063 - 77359691 on Build GRCh38 | Loss | KCNMA1 KCNMA1-AS3 |
nsv831919 | Chr.10:77292047 - 77470200 on Build GRCh38 | Loss | KCNMA1 KCNMA1-AS3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |