Assay Details
Target Gene Details
Entrez Gene ID: | 256536 |
Gene Name: | transcription elongation regulator 1 like |
Gene Aliases: |
- |
Location: |
Chr.10:131092391-131311721 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TCERG1L | NM_174937.3 | NP_777597.2 | ||
XM_011539577.1 | XP_011537879.1 | |||
AK096269.1 | ||||
BC042951.1 | AAH42951.1 | |||
BC093639.1 | AAH93639.1 | |||
BC101535.1 | AAI01536.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3237 | Chr.10:131110754 - 131114683 on Build GRCh38 | Loss |
![]() |
dgv182n67 | Chr.10:131110741 - 131114511 on Build GRCh38 | Loss |
![]() |
nsv832027 | Chr.10:131056320 - 131214052 on Build GRCh38 | Gain |
![]() ![]() |
esv6315 | Chr.10:131110545 - 131114773 on Build GRCh38 | Loss |
![]() |
esv26490 | Chr.10:131110462 - 131114883 on Build GRCh38 | Loss |
![]() |
nsv512193 | Chr.10:131110728 - 131114606 on Build GRCh38 | Loss |
![]() |
esv3547075 | Chr.10:131110688 - 131114898 on Build GRCh38 | Deletion |
![]() |
nsv1048253 | Chr.10:129847356 - 131188858 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1128533 | Chr.10:131109637 - 131115537 on Build GRCh38 | Deletion |
![]() |
nsv1054934 | Chr.10:130334322 - 133158679 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv7621 | Chr.10:131081871 - 131117163 on Build GRCh38 | Deletion |
![]() ![]() |
nsv470974 | Chr.10:131049527 - 131126683 on Build GRCh38 | Gain |
![]() ![]() |
nsv428242 | Chr.10:130607951 - 131583141 on Build GRCh38 | Gain |
![]() ![]() ![]() |
nsv511463 | Chr.10:131110205 - 131116646 on Build GRCh38 | Loss |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)