Assay Details
Target Gene Details
Entrez Gene ID: | 55844 |
Gene Name: | protein phosphatase 2 regulatory subunit Bdelta |
Gene Aliases: |
MDS026 |
Location: |
Chr.10:131900644-131971535 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PPP2R2D | XM_006717914.3 | XP_006717977.1 | ||
XM_017016407.1 | XP_016871896.1 | |||
AF220046.1 | AAF67639.1 | |||
AK000402.1 | ||||
AK293065.1 | ||||
BC047379.1 | AAH47379.1 | |||
BC058076.1 | AAH58076.1 | |||
BC072402.1 | AAH72402.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3624956 | Chr.10:131249138 - 132223150 on Build GRCh38 | Gain | BNIP3 TCERG1L LINC01164 JAKMIP3 PPP2R2D DPYSL4 STK32C |
nsv1054934 | Chr.10:130334322 - 133158679 on Build GRCh38 | Gain | LINC01164 NKX6-2 LINC01168 ADGRA1 LINC01166 PWWP2B BNIP3 LOC107984282 MIR378C LOC105378568 CFAP46 ADGRA1-AS1 TCERG1L INPP5A LRRC27 TCERG1L-AS1 JAKMIP3 C10orf91 PPP2R2D DPYSL4 LINC01167 STK32C |
nsv825650 | Chr.10:131914064 - 132056715 on Build GRCh38 | Gain | BNIP3 JAKMIP3 PPP2R2D |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |