Assay Details
Target Gene Details
Entrez Gene ID: | 643418 |
Gene Name: | lipase family member N |
Gene Aliases: |
ARCI8, LI4, LIPL4, bA186O14.3 |
Location: |
Chr.10:88757226-88783662 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LIPN | NM_001102469.1 | NP_001095939.1 | ||
XM_005270049.3 | XP_005270106.1 | |||
XM_011540083.2 | XP_011538385.1 | |||
XM_011540084.2 | XP_011538386.1 | |||
XM_011540085.2 | XP_011538387.1 | |||
XM_011540086.2 | XP_011538388.1 | |||
XM_017016549.1 | XP_016872038.1 | |||
EF426483.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv551840 | Chr.10:88718726 - 88841108 on Build GRCh38 | Gain | LIPM LIPN LIPK ANKRD22 |
nsv1043015 | Chr.10:88707310 - 88833935 on Build GRCh38 | Gain | LIPM LIPN LIPK ANKRD22 |
nsv1052613 | Chr.10:88740764 - 88766622 on Build GRCh38 | Loss | LIPN LIPK |
nsv831941 | Chr.10:88650731 - 88827575 on Build GRCh38 | Gain | LIPM LIPN LIPF LIPK ANKRD22 |
nsv825502 | Chr.10:88759237 - 88768756 on Build GRCh38 | Loss | LIPN |
esv3624139 | Chr.10:88724980 - 89003307 on Build GRCh38 | Gain | FAS-AS1 STAMBPL1 LIPM ACTA2 LIPN LIPK ACTA2-AS1 FAS ANKRD22 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |