Assay Details
Target Gene Details
Entrez Gene ID: | 664 |
Gene Name: | BCL2 interacting protein 3 |
Gene Aliases: |
NIP3 |
Location: |
Chr.10:131967683-131982013 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
BNIP3 | NM_004052.3 | NP_004043.3 | ||
AF002697.1 | AAC16738.1 | |||
AK222626.1 | BAD96346.1 | |||
AK295136.1 | ||||
BC021989.1 | AAH21989.1 | |||
BC067818.1 | AAH67818.1 | |||
BC080643.1 | AAH80643.1 | |||
HY049371.1 | ||||
U15174.1 | AAC00022.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3624956 | Chr.10:131249138 - 132223150 on Build GRCh38 | Gain | JAKMIP3 PPP2R2D BNIP3 TCERG1L DPYSL4 LINC01164 STK32C |
nsv1054934 | Chr.10:130334322 - 133158679 on Build GRCh38 | Gain | LOC107984282 LINC01166 INPP5A PWWP2B C10orf91 LINC01164 LOC105378568 NKX6-2 TCERG1L-AS1 LINC01167 JAKMIP3 PPP2R2D ADGRA1-AS1 BNIP3 TCERG1L DPYSL4 MIR378C LRRC27 ADGRA1 STK32C CFAP46 LINC01168 |
nsv825651 | Chr.10:131975920 - 131986071 on Build GRCh38 | Gain | BNIP3 |
nsv825650 | Chr.10:131914064 - 132056715 on Build GRCh38 | Gain | JAKMIP3 PPP2R2D BNIP3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |