Assay Details
Target Gene Details
Entrez Gene ID: | 119504 |
Gene Name: | anaphase promoting complex subunit 16 |
Gene Aliases: |
APC16, C10orf104, CENP-27, MSAG, bA570G20.3 |
Location: |
Chr.10:72216000-72235860 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ANAPC16 | NM_001242546.1 | NP_001229475.1 | ||
NM_001242547.1 | NP_001229476.1 | |||
NM_001242548.1 | NP_001229477.1 | |||
NM_173473.3 | NP_775744.1 | |||
NR_038391.1 | ||||
NR_038392.1 | ||||
BC009530.1 | AAH09530.1 | |||
BM768948.1 | ||||
CB984695.1 | ||||
CD511334.1 | ||||
DA310509.1 | ||||
DA739927.1 | ||||
EU552924.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv508592 | Chr.10:72164274 - 72227164 on Build GRCh38 | Deletion | ASCC1 ANAPC16 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |