Assay Details
Target Gene Details
Entrez Gene ID: | 64421 |
Gene Name: | DNA cross-link repair 1C |
Gene Aliases: |
A-SCID, DCLREC1C, RS-SCID, SCIDA, SNM1C |
Location: |
Chr.10:14897853-14954432 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1045188 | Chr.10:14936970 - 15015784 on Build GRCh38 | Loss | MEIG1 DCLRE1C LOC105376430 |
nsv1037136 | Chr.10:14941160 - 15018277 on Build GRCh38 | Gain+Loss | MEIG1 LOC105376432 DCLRE1C LOC105376430 |
nsv825256 | Chr.10:14944342 - 15027072 on Build GRCh38 | Gain | MEIG1 LOC105376432 DCLRE1C LOC105376430 |
nsv469720 | Chr.10:14865281 - 15037379 on Build GRCh38 | Gain | MEIG1 LOC105376432 DCLRE1C SUV39H2 HSPA14 LOC105376430 |
esv2733506 | Chr.10:14939255 - 15021366 on Build GRCh38 | Deletion | MEIG1 LOC105376432 DCLRE1C LOC105376430 |
nsv435909 | Chr.10:14940998 - 15022839 on Build GRCh38 | Deletion | MEIG1 LOC105376432 DCLRE1C LOC105376430 |
dgv68n27 | Chr.10:14913179 - 15018884 on Build GRCh38 | Gain | MEIG1 LOC105376432 DCLRE1C LOC105376430 |
dgv961n54 | Chr.10:14910793 - 15028616 on Build GRCh38 | Gain | MEIG1 LOC105376432 DCLRE1C LOC105376430 |
dgv178e212 | Chr.10:14936931 - 15024223 on Build GRCh38 | Gain | MEIG1 LOC105376432 DCLRE1C LOC105376430 |
dgv69n27 | Chr.10:14933172 - 15028616 on Build GRCh38 | Gain | MEIG1 LOC105376432 DCLRE1C LOC105376430 |
esv2760142 | Chr.10:14930788 - 15024489 on Build GRCh38 | Gain | MEIG1 LOC105376432 DCLRE1C LOC105376430 |
nsv831793 | Chr.10:14870604 - 15058763 on Build GRCh38 | Loss | MEIG1 LOC105376432 OLAH DCLRE1C SUV39H2 HSPA14 LOC105376430 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |