Assay Details
Target Gene Details
Entrez Gene ID: | 57705 |
Gene Name: | WDFY family member 4 |
Gene Aliases: |
C10orf64 |
Location: |
Chr.10:48684815-48982956 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
WDFY4 | NM_020945.1 | NP_065996.1 | ||
XM_005270004.3 | XP_005270061.1 | |||
XM_011539986.2 | XP_011538288.2 | |||
XM_011539987.2 | XP_011538289.2 | |||
XM_011539988.2 | XP_011538290.1 | |||
XM_011539990.2 | XP_011538292.1 | |||
XM_011539991.2 | XP_011538293.1 | |||
XM_011539992.2 | XP_011538294.1 | |||
XM_017016463.1 | XP_016871952.1 | |||
XM_017016465.1 | XP_016871954.1 | |||
AK127650.1 | BAC87073.1 | |||
BC034937.1 | AAH34937.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1042677 | Chr.10:47993784 - 49928329 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)