Assay Details
Target Gene Details
Entrez Gene ID: | 57178 |
Gene Name: | zinc finger MIZ-type containing 1 |
Gene Aliases: |
MIZ, RAI17, TRAFIP10, ZIMP10, hZIMP10 |
Location: |
Chr.10:79068994-79316528 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ZMIZ1 | NM_020338.3 | NP_065071.1 | ||
XM_005269988.3 | XP_005270045.1 | |||
XM_006717923.3 | XP_006717986.1 | |||
XM_006717924.3 | XP_006717987.1 | |||
XM_006717925.3 | XP_006717988.1 | |||
BC007288.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv551561 | Chr.10:78349364 - 79336832 on Build GRCh38 | Gain | ZMIZ1-AS1 ZMIZ1 |
nsv469807 | Chr.10:79043070 - 79212669 on Build GRCh38 | Loss | ZMIZ1-AS1 ZMIZ1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |