Assay Details
Target Gene Details
Entrez Gene ID: | 114299 |
Gene Name: | paralemmin 2 |
Gene Aliases: |
AKAP2 |
Location: |
Chr.9:109640788-109951476 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PALM2 | NM_001037293.2 | NP_001032370.1 | ||
AK095003.1 | BAC04472.1 | |||
BC039306.1 | AAH39306.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv509317 | Chr.9:109612244 - 109659231 on Build GRCh38 | Insertion | PALM2 |
nsv831686 | Chr.9:109599537 - 109770842 on Build GRCh38 | Gain | LOC107987013 PALM2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |