Assay Details
Target Gene Details
Entrez Gene ID: | 9550 |
Gene Name: | ATPase H+ transporting V1 subunit G1 |
Gene Aliases: |
ATP6G, ATP6G1, ATP6GL, ATP6J, Vma10 |
Location: |
Chr.9:114587714-114598872 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ATP6V1G1 | NM_004888.3 | NP_004879.1 | ||
AF038954.1 | AAC39868.1 | |||
BC008452.1 | AAH08452.1 | |||
BI596632.1 | ||||
CR456971.1 | CAG33252.1 | |||
CR542237.1 | CAG47033.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv948n27 | Chr.9:114193988 - 114693895 on Build GRCh38 | Gain | AKNA ORM1 LOC100505478 MIR455 TMEM268 WHRN ATP6V1G1 ORM2 COL27A1 |
nsv831694 | Chr.9:114566360 - 114740505 on Build GRCh38 | Gain | LOC100505478 TMEM268 ATP6V1G1 |
dgv7706n100 | Chr.9:114182745 - 114697870 on Build GRCh38 | Gain | AKNA ORM1 LOC100505478 MIR455 TMEM268 WHRN ATP6V1G1 ORM2 COL27A1 |
dgv33e196 | Chr.9:114030664 - 114604363 on Build GRCh38 | Duplication | AKNA KIF12 ORM1 MIR455 AMBP TMEM268 WHRN ZNF618 ATP6V1G1 ORM2 COL27A1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |