Assay Details
Target Gene Details
Entrez Gene ID: | 51162 |
Gene Name: | EGF like domain multiple 7 |
Gene Aliases: |
NEU1, VE-STATIN, ZNEU1 |
Location: |
Chr.9:136654753-136672678 on Build GRCh38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EGFL7 |
Target Gene Details
Entrez Gene ID: | 101928612 |
Gene Name: | uncharacterized LOC101928612 |
Gene Aliases: |
- |
Location: |
Chr.9:136648610-136660421 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HSPC324 | NR_135132.1 | |||
AF161442.1 | AAF29002.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv13005n54 | Chr.9:136605258 - 136696482 on Build GRCh38 | Loss | LINC01451 MIR126 AGPAT2 EGFL7 HSPC324 |
nsv509334 | Chr.9:136514168 - 136724663 on Build GRCh38 | Insertion | FAM69B LINC01451 MIR126 NOTCH1 MIR4674 AGPAT2 MIR4673 SNHG7 EGFL7 NALT1 HSPC324 |
nsv951203 | Chr.9:136562849 - 136770848 on Build GRCh38 | Deletion | SNORA17A MIR126 LCN10 LCN15 EGFL7 SNORA17B HSPC324 MIR6722 FAM69B LINC01451 LCN8 AGPAT2 SNHG7 LOC100128593 LCN6 |
nsv469918 | Chr.9:136359865 - 137173890 on Build GRCh38 | Loss | C9orf139 ABCA2 GRIN1 EDF1 LCN10 TMEM210 SNAPC4 SNORA17B FAM69B C8G MIR3621 NPDC1 AGPAT2 MIR4673 CCDC183 DPP7 LCN6 SDCCAG3 CARD9 MAN1B1 PMPCA PTGDS LCN12 FUT7 CLIC3 LOC100128593 TRAF2 SNORA17A MIR126 MIR4292 MAN1B1-AS1 UAP1L1 DNLZ TMEM141 HSPC324 C9orf172 NOTCH1 LCN8 SNHG7 RABL6 LRRC26 C9orf163 MAMDC4 SEC16A ENTPD2 PRR31 MIR4479 MIR4674 CCDC183-AS1 LCN15 EGFL7 FBXW5 MIR6722 LOC101930307 LINC01451 LCNL1 PHPT1 INPP5E C9orf142 SAPCD2 NALT1 |
esv2759720 | Chr.9:136356377 - 136932033 on Build GRCh38 | Loss | SNORA17A MIR126 MIR4292 EDF1 LCN10 SNAPC4 DNLZ TMEM141 SNORA17B HSPC324 FAM69B C9orf172 NOTCH1 LCN8 GPSM1 AGPAT2 MIR4673 SNHG7 CCDC183 RABL6 LCN6 C9orf163 MAMDC4 SDCCAG3 CARD9 SEC16A MIR4479 MIR4674 CCDC183-AS1 LCN15 EGFL7 PMPCA MIR6722 LINC01451 PHPT1 INPP5E LOC100128593 NALT1 TRAF2 |
nsv8579 | Chr.9:136505432 - 136760256 on Build GRCh38 | Gain | SNORA17A MIR126 LCN10 MIR4674 LCN15 EGFL7 SNORA17B HSPC324 MIR6722 FAM69B LINC01451 NOTCH1 LCN8 AGPAT2 MIR4673 SNHG7 LOC100128593 LCN6 NALT1 |
nsv615971 | Chr.9:136583078 - 136670216 on Build GRCh38 | Loss | LINC01451 EGFL7 HSPC324 |
nsv1053781 | Chr.9:136590694 - 136722927 on Build GRCh38 | Gain | FAM69B LINC01451 MIR126 AGPAT2 EGFL7 HSPC324 |
dgv957n27 | Chr.9:136619086 - 136725859 on Build GRCh38 | Loss | FAM69B MIR126 AGPAT2 SNHG7 EGFL7 HSPC324 |
esv2422220 | Chr.9:136584135 - 137350098 on Build GRCh38 | Deletion | C9orf139 NELFB ABCA2 SLC34A3 GRIN1 EDF1 LCN10 NDOR1 TMEM210 TUBB4B SSNA1 SNORA17B RNF208 FAM69B C8G MIR3621 NPDC1 AGPAT2 CCDC183 DPP7 LCN6 CYSRT1 MAN1B1 STPG3 RNF224 PTGDS LCN12 FUT7 CLIC3 LOC100128593 FAM166A TRAF2 TOR4A SNORA17A MIR126 MIR4292 MAN1B1-AS1 UAP1L1 TMEM141 HSPC324 C9orf172 TMEM203 LCN8 SNHG7 NRARP RABL6 LRRC26 MAMDC4 ENTPD2 PRR31 MIR4479 CCDC183-AS1 LCN15 EXD3 EGFL7 FBXW5 MIR6722 LOC101930307 LINC01451 LCNL1 PHPT1 TPRN C9orf142 SAPCD2 ANAPC2 C9orf173-AS1 |