Assay Details
Target Gene Details
Entrez Gene ID: | 1271 |
Gene Name: | ciliary neurotrophic factor receptor |
Gene Aliases: |
- |
Location: |
Chr.9:34551432-34590386 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CNTFR | NM_001207011.1 | NP_001193940.1 | ||
NM_001842.4 | NP_001833.1 | |||
NM_147164.2 | NP_671693.1 | |||
XM_017014260.1 | XP_016869749.1 | |||
XM_017014261.1 | XP_016869750.1 | |||
XM_017014262.1 | XP_016869751.1 | |||
XM_017014263.1 | XP_016869752.1 | |||
XM_017014264.1 | XP_016869753.1 | |||
XM_017014265.1 | XP_016869754.1 | |||
AK127444.1 | ||||
BC001492.2 | AAH01492.1 | |||
BT019824.1 | AAV38627.1 | |||
HQ628632.1 | ||||
M73238.1 | AAA35707.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3620335 | Chr.9:33871811 - 34738198 on Build GRCh38 | Gain | C9orf24 UBAP1 CCL21 KIAA1161 CCL27 KIF24 GALT IL11RA DCAF12 UBAP2 ARID3C RPP25L FAM205A CNTFR SNORD121A LOC114224 CNTFR-AS1 SNORD121B DCTN3 CCL19 LOC730098 FAM219A DNAI1 NUDT2 ENHO UBE2R2 SIGMAR1 |
nsv1161857 | Chr.9:34537279 - 34669174 on Build GRCh38 | Deletion | RPP25L CNTFR CNTFR-AS1 SIGMAR1 CCL27 DCTN3 GALT ARID3C LOC730098 IL11RA |
dgv12659n54 | Chr.9:34552439 - 34663141 on Build GRCh38 | Loss | RPP25L CNTFR CNTFR-AS1 SIGMAR1 CCL27 DCTN3 GALT ARID3C IL11RA |
nsv831543 | Chr.9:34431291 - 34615384 on Build GRCh38 | Loss | RPP25L CNTFR ENHO CNTFR-AS1 DCTN3 FAM219A DNAI1 |
nsv951699 | Chr.9:34553903 - 34574802 on Build GRCh38 | Deletion | CNTFR CNTFR-AS1 |
nsv818699 | Chr.9:34535211 - 34685943 on Build GRCh38 | Gain | RPP25L CNTFR CNTFR-AS1 SIGMAR1 CCL27 DCTN3 GALT ARID3C LOC730098 IL11RA |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |