Assay Details
Target Gene Details
Entrez Gene ID: | 10783 |
Gene Name: | NIMA related kinase 6 |
Gene Aliases: |
SID6-1512 |
Location: |
Chr.9:124257606-124352442 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NEK6 | NM_001145001.2 | NP_001138473.1 | ||
NM_001166167.1 | NP_001159639.1 | |||
NM_001166168.1 | NP_001159640.1 | |||
NM_014397.5 | NP_055212.2 | |||
XM_005251664.1 | XP_005251721.1 | |||
AB026289.1 | BAA85045.1 | |||
AF087909.1 | AAG13417.1 | |||
AK313071.1 | ||||
BC000101.2 | AAH00101.2 | |||
BC004174.2 | AAH04174.2 | |||
BC004209.2 | AAH04209.2 | |||
BC012761.2 | AAH12761.1 | |||
BM083872.1 | ||||
BP265230.1 | ||||
BP279761.1 | ||||
CF994196.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831719 | Chr.9:124093337 - 124279941 on Build GRCh38 | Gain | LOC613206 NEK6 |
nsv519767 | Chr.9:124247983 - 124364893 on Build GRCh38 | Loss | PSMB7 LOC100129034 LOC613206 NEK6 |
nsv471322 | Chr.9:124247835 - 124357163 on Build GRCh38 | Loss | PSMB7 LOC100129034 LOC613206 NEK6 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |