Assay Details
Target Gene Details
Entrez Gene ID: | 29882 |
Gene Name: | anaphase promoting complex subunit 2 |
Gene Aliases: |
APC2 |
Location: |
Chr.9:137174784-137188605 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ANAPC2 | NM_013366.3 | NP_037498.1 | ||
AB037827.1 | BAA92644.1 | |||
AF191337.1 | AAF05751.1 | |||
AK294601.1 | ||||
BC001579.1 | AAH01579.1 | |||
BC009487.2 | AAH09487.2 | |||
BC032503.1 | AAH32503.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv616007 | Chr.9:137079368 - 137240666 on Build GRCh38 | Loss | GRIN1 LOC101930307 SSNA1 MIR3621 CYSRT1 UAP1L1 MAN1B1-AS1 RNF208 RNF224 NDOR1 SLC34A3 DPP7 LRRC26 MAN1B1 TMEM203 TMEM210 TPRN ANAPC2 |
dgv13009n54 | Chr.9:137173890 - 137327859 on Build GRCh38 | Loss | SSNA1 CYSRT1 TOR4A STPG3 NRARP RNF208 RNF224 NDOR1 C9orf173-AS1 SLC34A3 EXD3 TUBB4B TMEM203 FAM166A NELFB TPRN ANAPC2 |
nsv524322 | Chr.9:136795692 - 137482216 on Build GRCh38 | Loss | GRIN1 LCNL1 FUT7 NOXA1 ENTPD2 UAP1L1 C9orf139 NRARP RNF208 SAPCD2 TMEM203 C9orf142 ANAPC2 C8G CYSRT1 TOR4A NPDC1 NDOR1 C9orf173-AS1 MAMDC4 RABL6 NELFB TMEM210 PNPLA7 TPRN PRR31 NSMF SSNA1 MIR3621 PTGDS MAN1B1-AS1 MIR7114 PHPT1 C9orf172 TRAF2 FBXW5 MIR4292 LCN12 LOC101930307 EDF1 ENTPD8 STPG3 CCDC183 CLIC3 RNF224 SLC34A3 EXD3 DPP7 LRRC26 TUBB4B MAN1B1 MIR4479 ABCA2 FAM166A CCDC183-AS1 |
nsv951207 | Chr.9:137136949 - 137379148 on Build GRCh38 | Deletion | GRIN1 SSNA1 MIR3621 CYSRT1 TOR4A STPG3 NRARP RNF208 RNF224 NDOR1 C9orf173-AS1 SLC34A3 EXD3 LRRC26 TUBB4B TMEM203 FAM166A NELFB TMEM210 TPRN ANAPC2 |
nsv1161918 | Chr.9:137118319 - 137438026 on Build GRCh38 | Duplication | GRIN1 NOXA1 SSNA1 MIR3621 CYSRT1 TOR4A ENTPD8 STPG3 NRARP RNF208 RNF224 NDOR1 C9orf173-AS1 SLC34A3 EXD3 LRRC26 TUBB4B TMEM203 FAM166A NELFB TMEM210 TPRN ANAPC2 |
esv2761561 | Chr.9:137030185 - 137191593 on Build GRCh38 | Gain | GRIN1 FUT7 LOC101930307 SSNA1 MIR3621 ENTPD2 UAP1L1 C9orf139 NPDC1 MAN1B1-AS1 SAPCD2 DPP7 LRRC26 MAN1B1 TMEM210 ANAPC2 |
esv2422220 | Chr.9:136584135 - 137350098 on Build GRCh38 | Deletion | LCN15 GRIN1 LCNL1 FUT7 EGFL7 ENTPD2 UAP1L1 C9orf139 NRARP RNF208 SAPCD2 TMEM203 C9orf142 ANAPC2 C8G FAM69B TMEM141 CYSRT1 TOR4A SNORA17A NPDC1 HSPC324 NDOR1 C9orf173-AS1 MAMDC4 RABL6 NELFB TMEM210 TPRN AGPAT2 PRR31 LINC01451 LCN6 SSNA1 MIR3621 LCN8 PTGDS MAN1B1-AS1 PHPT1 C9orf172 TRAF2 FBXW5 MIR4292 LCN12 MIR126 SNHG7 LCN10 LOC101930307 EDF1 LOC100128593 SNORA17B STPG3 CCDC183 CLIC3 RNF224 SLC34A3 EXD3 DPP7 LRRC26 TUBB4B MAN1B1 MIR4479 ABCA2 FAM166A MIR6722 CCDC183-AS1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |